Learn More
OHAUS™ Holder Clamp
OHAUS™ Holder Clamp provides secure support for holding glassware at elevated positions.
Brand:  OHAUS™ 30392265
 View more versions of this product
Product Code. 15792309
  • £10.35 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel is autoclavable and corrosion-resistant
  • Aluminum is lightweight yet strong
  • Cast iron with black enamel finish is chemical-resistant
  • Available in different shapes and sizes to fit your laboratory needs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Holder
Clamp, Holder, Universal, CLC-UNSMBA
0 to 22 mm
Aluminum
Universal