missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ Holder Clamp for Fiber Optic Light Guides or LED Ring Lights

Holder Clamp to position fiber optic light guides or LED Ring Lights precisely. For use with an articulating arm (158345) and base (158 340).

Brand:  SCHOTT AG Lighting and Imaging™ 158330

Additional Details : Weight : 5.00000kg

 View more versions of this product

Product Code. 15793022

  • £76.55 / 1 unit
Estimated Shipment: 27-09-2024
to see stock.
Add to basket

Description

Description

  • Holder clamp to position fiber optic flexible light guides or Ring Lights precisely.
  • 1 x M6 hole.
  • Articulating arm and base available separately.
  • Holders for different size light guides and focusing lenses.
Specifications

Specifications

Holder Clamp
5 mm
Stereo Microscopy Illumination
1 M6 hole
0.2 in.
40 x 18 x 11 mm
Flexible Lightguides up to 5 mm dia.
Articulating arm 158 345 and optional a base 158 340
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers