missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Optifit Twin™ Full-Face Air Purifying Respirator (APR)

The Optifit Twin™ Full-Face APR provides protection versus particulates and/or toxic gases. It features a full lens for wide field of vision, a silicone body for optimum fit, and a click-fit cartridge system for flexible protection. A built-in diaphragm allows for better speech transmission.

Brand:  Honeywell™ 1715241

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11856930

  • £278.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Product Type: Full-Face Air Purifying Respirator (APR)
  • Description: Polycarbonate Visor with Silicone Skirt, Face Blank, and Exhalation Valve. Integrated Plastic Diaphragm
  • Cartridge/Filter: Bayonet N-Series Filter (EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters Only)
  • Lens Type: Full Lens
  • Material (Lens): Polycarbonate
  • Harness Style: Five-Strap Head Harness, with Quick Release Adjusting Buckles
  • Certifications/Compliance: EN136:1998, CE PPE Cat:3, QA: ISO 9001/2000
  • For Use With (Application): Laboratories, Pharmaceutical Industry, Oil and Gas Industry, Mechanical Industry, Automotive Industry, Chemical Industry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bayonet N-Series filter (EN 141 or EN 143 Class 1 or 2 cert Honeywell filters only)
EN 136:1998, EC Category PPE: 3, QA: ISO 9001/2000, EC Certificate: 0070-236-079-06-06-0023, EC Attestation: 0070-236-079-06-06-0023, Edeclaration of conformity INRS - 1715231
Full-Face Air Purifying Respirator (APR)
Yes
Laboratories, Pharmaceutical Industry, Oil and Gas Industry, Mechanical Industry, Automotive Industry, Chemical Industry
Silicone Skirt, Filter, Diaphragm
Yes
570 g
Double filter
Black
Medium
Silicone
Five-strap head harness, with quick release adjusting buckles
Polycarbonate
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.