Learn More
Scientific Industries SI™ Horizontal Tube Holder
Snap tubes into place swiftly and easily. Scientific Instruments SI™ Horizontal Tube Holder does not require additional attachments for use.
£107.00 - £240.00
Specifications
For Use With (Equipment) | Vortex-Genie 2 Mixer and the Vortex-Genie Pulse |
---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15507355
|
Scientific Industries SI™
SI-H506 |
50 mL Tubes |
£107.00
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15517355
|
Scientific Industries SI™
SI-H512 |
12 x 15 mL Tubes |
£177.00
Each |
In Stock
Log in to see stock. |
|||||
15527355
|
Scientific Industries SI™
SI-H524 |
24 Microtubes |
£240.00
Each |
In Stock
Log in to see stock. |
|||||
Description
- Holds tubes in place horizontally.
- Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
- No additional attachments are necessary
- Holds tubes in place horizontally.
- Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
- No additional attachments are necessary
Specifications
Vortex-Genie 2 Mixer and the Vortex-Genie Pulse |