missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT HPLC Suction Filters
Suction filters for use with HPLC instruments.
Brand: SCAT 300022
62.56 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Be prepared for HPLC with these SCAT™ suction filters.
Specifications
10 μm | |
3.2 mm | |
PFTE, PFA |
HPLC Suction Filter | |
5 Pk. | |
HPLC Instrument |