Learn More
SCAT HPLC Supply-Sets, V2.0
The HPLC Supply Sets, V2.0 of the manufacturer SCAT are combinations of equipment for contamination-free supply of HPLC systems with solvents. The sets consist of a laboratory bottle with GL 45 thread, a SCAT SafetyCap as well as capillaries, suction filters and air valves.
Brand:  SCAT™ 307327
107.62 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.58680kg
 View more versions of this product
Product Code. 17927260
  • £141.10 / 1 set
Estimated Shipment: 18-07-2024
to see stock.
Description
Description
  • Customised system with high quality set components
  • SafetyCaps of the premium manufacturer SCAT
  • Laboratory bottles
  • Price attractive in comparison with single purchase
  • Ready for use, no further components necessary
SCAT offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
Specifications
Specifications
1 x SafetyCap II GL28, 1x 100 mL Laboratory bottle round, brown glass, 2 x 1.5 m capillaries 3.2 mm OD, 2 x PTFE suction filters, air valve (317010)
Complete Solvent Collection System
PTFE
GL45
Solvent supply for your HPLC
Amber
Any HPLC System