missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ HT ExoSAP-IT™ High-Throughput PCR Product Cleanup

HT ExoSAP-IT High-Throughput PCR Product Clean-up is an alternative formulation of ExoSAP-IT reagent specifically designed for the unique requirements of high-throughput, automated platforms, and multi-channel pipettes.

Brand:  Applied Biosystems™ 783951000RX

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 15827121

  • £846.00 / 1000 reactions
Estimated Shipment: 09-08-2024
to see stock.

Description

Description

  • One-tube PCR clean-up - Add HT ExoSAP-IT reagent directly to PCR product
  • Exceptional accuracy - Achieve high quality data even with long read lengths
  • 100% sample recovery - No loss of PCR products regardless of the fragment size
  • Simple, single-step - Replaces the multiple steps and wasted time inherent with beads or columns
  • High-throughput processing - Even faster time to results with a low viscosity formulation, allowing robotic pipetting
  • Scalable - Treat reaction volumes from 5μL to 5L
  • Convenient packaging - Available in 8-tube strips and 12 strips in a 96-well plate
  • Stable at 25°C for 8 hours - Retains full functional activity and at 45°C is stable for one week
Specifications

Specifications

45°C (One Week)
Sequencing
PCR Product Clean-up
PCR Product Clean-up, 1000 rxns, Single Step, High-throughput Processing, Scalable, Specifically Designed for the Unique Requirements of High-throughput, Automated Platforms and Multi-channel Pipettes
1000
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.