missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ HT ExoSAP-IT™ High-Throughput PCR Product Cleanup
HT ExoSAP-IT High-Throughput PCR Product Clean-up is an alternative formulation of ExoSAP-IT reagent specifically designed for the unique requirements of high-throughput, automated platforms, and multi-channel pipettes.
£429.00 - £3550.00
Specifications
Content And Storage | 45°C (One Week) |
---|---|
Product Type | PCR Product Clean-up |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15837121
|
Applied Biosystems™
783951EA |
480 Reactions |
£429.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15827121
|
Applied Biosystems™
783951000RX |
1000 |
£846.00
1000 reactions |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15809916
|
Applied Biosystems™
783955000RX |
5000 Reactions |
£3550.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899906
|
Applied Biosystems™
783951000RX |
1000 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- One-tube PCR clean-up - Add HT ExoSAP-IT reagent directly to PCR product
- Exceptional accuracy - Achieve high quality data even with long read lengths
- 100% sample recovery - No loss of PCR products regardless of the fragment size
- Simple, single-step - Replaces the multiple steps and wasted time inherent with beads or columns
- High-throughput processing - Even faster time to results with a low viscosity formulation, allowing robotic pipetting
- Scalable - Treat reaction volumes from 5μL to 5L
- Convenient packaging - Available in 8-tube strips and 12 strips in a 96-well plate
- Stable at 25°C for 8 hours - Retains full functional activity and at 45°C is stable for one week
Specifications
45°C (One Week) | |
PCR Product Clean-up |
For Research Use Only. Not for use in diagnostic procedures.