Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications. Ideal for high-throughput PCR product cleanup using liquid handling platforms and multi-channel pipettes.
Brand: Applied Biosystems™
606.69 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.47500kg
Description
- Mixture of Exonuclease I and Shrimp Alkaline Phosphatase for the enzymatic removal of excess primers and dNTPs following a PCR reaction
- One-tube PCR cleanup
- Shortened 14-minute protocol
- Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
Specifications
Store at −20°C. Stable for one month at 4°C | |
1000 | |
2 mL |