Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications. Ideal for high-throughput PCR product cleanup using liquid handling platforms and multi-channel pipettes.
£465.00 - £18420.00
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15553687
|
Applied Biosystems™
785951EA |
480 reactions, 8-tube strip |
£465.00
480 reactions |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15563687
|
Applied Biosystems™
785951PK |
5,760 reactions, 12 × 8-tube strips in a tray |
£5210.00
5760 reactions |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15583687
|
Applied Biosystems™
785951000RXN |
1000 |
£910.00
1000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15503697
|
Applied Biosystems™
785954PK |
23,040 reactions, 48 × 8-tube strips in a tray |
£18420.00
23040 reactions |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15513697
|
Applied Biosystems™
785955000RXN |
5000 |
£3825.00
5000 reactions |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Mixture of Exonuclease I and Shrimp Alkaline Phosphatase for the enzymatic removal of excess primers and dNTPs following a PCR reaction
- One-tube PCR cleanup
- Shortened 14-minute protocol
- Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
Specifications
Store at −20°C. Stable for one month at 4°C |