Learn More
Human Desmoplakin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB9080-SP
Description
Desmoplakin Monoclonal specifically detects Desmoplakin in Human samples. It is validated for Western Blot.
Specifications
Desmoplakin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
250/210 kDa paraneoplastic pemphigus antigen, DCWHKTA, desmoplakin, desmoplakin (DPI, DPII), desmoplakin I, desmoplakin II, DP, DPI, DPII, DSP, KPPS2, PPKS2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1832 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot | |
824038 | |
Western Blot 0.5 ug/mL | |
P15924 | |
DSP | |
E. coli-derived recombinant human Desmoplakin Asn1796-Val2046 Accession # P15924 | |
25 μg | |
Primary | |
Detects human Desmoplakin in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only