missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ECM-1 ELISA Kit
Human ECM-1 quantitates human ECM-1 in serum, plasma, supernatant.
Brand: Invitrogen™ EH165RBX10
This item is not returnable.
View return policy
Description
Human ECM-1 quantitates human ECM-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ECM-1.
Secretory Component is an epithelial transport receptor for polymeric IgA and IgM. Antibody to secretory component has been shown to be useful for identifying glandular carcinomas.Specifications
AI663821; ECM1; extracellular matrix protein 1; p85; RP11-54A4.6; Secretory component p85; testicular tissue protein Li 61; URBWD | |
24 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
ECM1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
24 to 6,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q16610 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
ECM1 | |
4 hr 45 min |