missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ECM-1 ELISA Kit
Human ECM-1 quantitates human ECM-1 in serum, plasma, supernatant.
£540.00 - £3870.00
Specifications
Accession Number | AI663821; ECM1; extracellular matrix protein 1; p85; RP11-54A4.6; Secretory component p85; testicular tissue protein Li 61; URBWD |
---|---|
Assay Range | 24 to 6,000 pg/mL |
Assay Sensitivity | 24 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17124919
|
Invitrogen™
EH165RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
17834080
|
Invitrogen™
EH165RBX10 |
10 x 96 Tests |
£3870.00
Pack of 10 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
17174443
|
Invitrogen™
EH165RB |
96 Tests |
£540.00
Pack of 96 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
Description
Human ECM-1 quantitates human ECM-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ECM-1.
Secretory Component is an epithelial transport receptor for polymeric IgA and IgM. Antibody to secretory component has been shown to be useful for identifying glandular carcinomas.Specifications
AI663821; ECM1; extracellular matrix protein 1; p85; RP11-54A4.6; Secretory component p85; testicular tissue protein Li 61; URBWD | |
24 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
ECM1 | |
<10% | |
HRP | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
ECM1 | |
4 hr 45 min |
24 to 6,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q16610 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |