missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-20 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 100-41-500UG
This item is not returnable.
View return policy
Description
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.Specifications
Q9NP95 | |
protein with no preservative | |
23.2 | |
500 μg | |
-20°C | |
<1 EU/μg | |
FGF20 | |
Unconjugated | |
PLAEVGGFLG GLEGLGQQVG SHFLLPPAGE RPPLLGERRS AAERSARGGP GAAQLAHLHG ILRRRQLYCR TGFHLQILPD GSVQGTRQDH SLFGILEFIS VAVGLVSIRG VDSGLYLGMN DKGELYGSEK LTSECIFREQ FEENWYNTYS SNIYKHGDTG RRYFVALNKD GTPRDGARSK RHQKFTHFLP RPVDPERVPE LYKDLLMYT | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
26281 | |
Human FGF-20 | |
E. coli | |
RUO | |
FGF; Fgf20; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20; RHDA2 | |
Human | |
Recombinant | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction