missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-20 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £3125.00
Specifications
Accession Number | Q9NP95 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 26281 |
Molecular Weight (g/mol) | 23.2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17851433
|
Gibco™
100-41-100UG |
100 μg |
£624.00
100µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17800853
|
Gibco™
100-41-15UG |
15 μg |
£158.00
15µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17840263
|
Gibco™
100-41-1MG |
2 x 500 μg |
£3125.00
1mg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17800373
|
Gibco™
100-41-250UG |
250 μg |
£1094.00
250µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17850093
|
Gibco™
100-41-3UG |
3 μg |
£65.00
3µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17870653
|
Gibco™
100-41-500UG |
500 μg |
£1876.00
500µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.Specifications
Q9NP95 | |
protein with no preservative | |
23.2 | |
E. coli | |
RUO | |
FGF; Fgf20; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20; RHDA2 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
26281 | |
Human FGF-20 | |
-20°C | |
<1 EU/μg | |
FGF20 | |
Unconjugated | |
PLAEVGGFLG GLEGLGQQVG SHFLLPPAGE RPPLLGERRS AAERSARGGP GAAQLAHLHG ILRRRQLYCR TGFHLQILPD GSVQGTRQDH SLFGILEFIS VAVGLVSIRG VDSGLYLGMN DKGELYGSEK LTSECIFREQ FEENWYNTYS SNIYKHGDTG RRYFVALNKD GTPRDGARSK RHQKFTHFLP RPVDPERVPE LYKDLLMYT | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |