Learn More
Gibco™ Human FGF-20 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 100-41-01M
 View more versions of this product
Product Code. 30088142
  • £3125.00 / Pack of 2
Explore more special offers
Description
Description
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q9NP95
Protein with no preservative
23.2
2 x 500 μg
-20°C
<1 EU/μg
FGF20
Unconjugated
PLAEVGGFLG GLEGLGQQVG SHFLLPPAGE RPPLLGERRS AAERSARGGP GAAQLAHLHG ILRRRQLYCR TGFHLQILPD GSVQGTRQDH SLFGILEFIS VAVGLVSIRG VDSGLYLGMN DKGELYGSEK LTSECIFREQ FEENWYNTYS SNIYKHGDTG RRYFVALNKD GTPRDGARSK RHQKFTHFLP RPVDPERVPE LYKDLLMYT
≥ 90% by SDS-PAGE gel and HPLC analyses.
Functional Assay
26281
Human FGF-20
E. coli
RUO
FGF; Fgf20; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20; RHDA2
Human
Recombinant
Lyophilized