missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-5 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 100-34-10UG
58.50 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Plays an important role in the regulation of cell proliferation and cell differentiation. Required for normal regulation of the hair growth cycle. Functions as an inhibitor of hair elongation by promoting progression from anagen, the growth phase of the hair follicle, into catagen the apoptosis-induced regression phase.Specifications
P12034 | |
protein with no preservative | |
27.6 | |
10 μg | |
-20°C | |
<1 EU/μg | |
FGF5 | |
Unconjugated | |
MAWAHGEKRL APKGQPGPAA TDRNPIGSSS RQSSSSAMSS SSASSSPAAS LGSQGSGLEQ SSFQWSPSGR RTGSLYCRVG IGFHLQIYPD GKVNGSHEAN MLSVLEIFAV SQGIVGIRGV FSNKFLAMSK KGKLHASAKF TDDCKFRERF QENSYNTYAS AIHRTEKTGR EWYVALNKRG KAKRGCSPRV KPQHISTHFL PRFKQSEQPE LSFTVTVPEK KNPPSPIKSK IPLSAPRKNT NSVKYRLKFR FG | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
2250 | |
Human FGF-5 | |
E. coli | |
RUO | |
angora; FGF; FGF5; FGF-5; Fibroblast growth factor; Fibroblast growth factor 5; fibroblast growth factor FGF-5; go; HBGF-5; Heparin-binding growth factor 5; Smag-82; TCMGLY | |
Human | |
Recombinant | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction