missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-5 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £1206.00
Specifications
Accession Number | P12034 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2250 |
Molecular Weight (g/mol) | 27.6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17881653
|
Gibco™
100-34-100UG |
100 μg |
£241.00
100µg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17870443
|
Gibco™
100-34-10UG |
10 μg |
£65.00
10µg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17890173
|
Gibco™
100-34-1MG |
1 mg |
£1206.00
1mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17840933
|
Gibco™
100-34-250UG |
250 μg |
£423.00
250µg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17820923
|
Gibco™
100-34-500UG |
500 μg |
£724.00
500µg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17821433
|
Gibco™
100-34-50UG |
50 μg |
£158.00
50µg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Plays an important role in the regulation of cell proliferation and cell differentiation. Required for normal regulation of the hair growth cycle. Functions as an inhibitor of hair elongation by promoting progression from anagen, the growth phase of the hair follicle, into catagen the apoptosis-induced regression phase.Specifications
P12034 | |
protein with no preservative | |
27.6 | |
E. coli | |
RUO | |
angora; FGF; FGF5; FGF-5; Fibroblast growth factor; Fibroblast growth factor 5; fibroblast growth factor FGF-5; go; HBGF-5; Heparin-binding growth factor 5; Smag-82; TCMGLY | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
2250 | |
Human FGF-5 | |
-20°C | |
<1 EU/μg | |
FGF5 | |
Unconjugated | |
MAWAHGEKRL APKGQPGPAA TDRNPIGSSS RQSSSSAMSS SSASSSPAAS LGSQGSGLEQ SSFQWSPSGR RTGSLYCRVG IGFHLQIYPD GKVNGSHEAN MLSVLEIFAV SQGIVGIRGV FSNKFLAMSK KGKLHASAKF TDDCKFRERF QENSYNTYAS AIHRTEKTGR EWYVALNKRG KAKRGCSPRV KPQHISTHFL PRFKQSEQPE LSFTVTVPEK KNPPSPIKSK IPLSAPRKNT NSVKYRLKFR FG | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |