missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-7 (KGF), Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
Brand: Gibco™ AF-100-19-500UG
This item is not returnable.
View return policy
Description
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Specifications
P21781 | |
Lyophilized | |
2252 | |
FGF7 | |
500 μg | |
RUO | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
Protein | |
Unconjugated | |
CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT | |
E. coli |
Functional Assay | |
protein with no preservative | |
18.9 kDa | |
HPLC, SDS-PAGE | |
-20°C | |
<0.1 EU/ μg | |
FGF7 | |
Human | |
Recombinant | |
Determined by a cell proliferation assay using 4 MBr-5 cells. The expected ED50 for this effect is 0.1-15 ng/ml | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |