missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-7 (KGF), Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
£77.00 - £2653.00
Specifications
Accession Number | P21781 |
---|---|
For Use With (Application) | Functional Assay |
Format | Lyophilized |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2252 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17806888
|
Gibco™
AF-100-19-2UG |
2 μg |
£77.00
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17876418
|
Gibco™
AF-100-19-50UG |
50 μg |
£566.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17846438
|
Gibco™
AF-100-19-500UG |
500 μg |
£2653.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17836528
|
Gibco™
AF-100-19-250UG |
250 μg |
£1592.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Specifications
P21781 | |
Lyophilized | |
2252 | |
FGF7 | |
-20°C | |
<0.1 EU/ μg | |
FGF7 | |
Human | |
Recombinant | |
Determined by a cell proliferation assay using 4 MBr-5 cells. The expected ED50 for this effect is 0.1-15 ng/ml | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
protein with no preservative | |
18.9 kDa | |
HPLC, SDS-PAGE | |
RUO | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
Protein | |
Unconjugated | |
CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT | |
E. coli |