missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FRP-1 (soluble) Recombinant Protein, PeproTech®
HeLa cells Recombinant Protein
Brand: Gibco™ 120-29-100UG
This item is not returnable.
View return policy
Description
Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.Specifications
Q8N474 | |
protein with no preservative | |
32.5 | |
100 μg | |
-20°C | |
<1 EU/μg | |
SFRP1 | |
Unconjugated | |
SEYDYVSFQS DIGPYQSGRF YTKPPQCVDI PADLRLCHNV GYKKMVLPNL LEHETMAEVK QQASSWVPLL NKNCHAGTQV FLCSLFAPVC LDRPIYPCRW LCEAVRDSCE PVMQFFGFYW PEMLKCDKFP EGDVCIAMTP PNATEASKPQ GTTVCPPCDN ELKSEAIIEH LCASEFALRM KIKEVKKENG DKKIVPKKKK PLKLGPIKKK DLKKLVLYLK NGADCPCHQL DNLSHHFLIM GRKVKSQYLL TAIHKWDKKN KEFKNFMKKM KNHECPTFQS VFK | |
≥ 97% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
6422 | |
Human FRP-1 (soluble) | |
HeLa cells | |
RUO | |
2210415K03Rik; AW011917; AW107218; AW742929; frizzled-related protein; FRP; FRP1; FRP-1; FrzA; SARP2; SARP-2; Secreted apoptosis-related protein 2; secreted frizzled related protein 1; secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; SFRP1; sFRP-1 | |
Human | |
Recombinant | |
Lyophilized |