missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FRP-1 (soluble) Recombinant Protein, PeproTech®
HeLa cells Recombinant Protein
£65.00 - £3125.00
Specifications
Accession Number | Q8N474 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 6422 |
Molecular Weight (g/mol) | 32.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17820303
|
Gibco™
120-29-100UG |
100 μg |
£624.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17841263
|
Gibco™
120-29-1MG |
1 mg |
£3125.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861473
|
Gibco™
120-29-20UG |
20 μg |
£158.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17859922
|
Gibco™
120-29-250UG |
250 μg |
£1094.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830973
|
Gibco™
120-29-500UG |
500 μg |
£1876.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17880123
|
Gibco™
120-29-5UG |
5 μg |
£65.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.Specifications
Q8N474 | |
protein with no preservative | |
32.5 | |
HeLa cells | |
RUO | |
2210415K03Rik; AW011917; AW107218; AW742929; frizzled-related protein; FRP; FRP1; FRP-1; FrzA; SARP2; SARP-2; Secreted apoptosis-related protein 2; secreted frizzled related protein 1; secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; SFRP1; sFRP-1 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
6422 | |
Human FRP-1 (soluble) | |
-20°C | |
<1 EU/μg | |
SFRP1 | |
Unconjugated | |
SEYDYVSFQS DIGPYQSGRF YTKPPQCVDI PADLRLCHNV GYKKMVLPNL LEHETMAEVK QQASSWVPLL NKNCHAGTQV FLCSLFAPVC LDRPIYPCRW LCEAVRDSCE PVMQFFGFYW PEMLKCDKFP EGDVCIAMTP PNATEASKPQ GTTVCPPCDN ELKSEAIIEH LCASEFALRM KIKEVKKENG DKKIVPKKKK PLKLGPIKKK DLKKLVLYLK NGADCPCHQL DNLSHHFLIM GRKVKSQYLL TAIHKWDKKN KEFKNFMKKM KNHECPTFQS VFK | |
≥ 97% by SDS-PAGE gel and HPLC analyses. |