missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GDF-5 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 120-01-250UG
This item is not returnable.
View return policy
Description
GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Hunter-Thompson type dwarfism and in Grebe Syndrome, which is characterized by short stature, extra digits, and short and deformed extremities. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino acid signal peptide and a 354 amino acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.Specifications
P43026 | |
protein with no preservative | |
27 | |
250 μg | |
-20°C | |
<1 EU/μg | |
GDF5 | |
Unconjugated | |
APLATRQGKR PSKNLKARCS RKALHVNFKD MGWDDWIIAP LEYEAFHCEG LCEFPLRSHL EPTNHAVIQT LMNSMDPEST PPTCCVPTRL SPISILFIDS ANNVVYKQYE DMVVESCGCR | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
8200 | |
Human GDF-5 | |
E. coli | |
RUO | |
BDA1C; Bmp14; BMP-14; Bone morphogenetic protein 14; bp; brachypodism; brp; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP1; Cdmp-1; GDF5; GDF-5; growth differentiation factor 5; growth/differentiation factor 5; H-GDF-5; LAP4; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; OS5; radotermin; SYM1B; SYNS2 | |
Human | |
Recombinant | |
Lyophilized |