missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GDF-5 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £1200.00
Specifications
Accession Number | P43026 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 8200 |
Molecular Weight (g/mol) | 27 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17841323
|
Gibco™
120-01-100UG |
100 μg |
£240.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17850773
|
Gibco™
120-01-10UG |
10 μg |
£64.75
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17831733
|
Gibco™
120-01-1MG |
1 mg |
£1200.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17891243
|
Gibco™
120-01-250UG |
250 μg |
£421.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17880743
|
Gibco™
120-01-500UG |
500 μg |
£720.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870373
|
Gibco™
120-01-50UG |
50 μg |
£157.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Hunter-Thompson type dwarfism and in Grebe Syndrome, which is characterized by short stature, extra digits, and short and deformed extremities. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino acid signal peptide and a 354 amino acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.Specifications
P43026 | |
protein with no preservative | |
27 | |
E. coli | |
RUO | |
BDA1C; Bmp14; BMP-14; Bone morphogenetic protein 14; bp; brachypodism; brp; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP1; Cdmp-1; GDF5; GDF-5; growth differentiation factor 5; growth/differentiation factor 5; H-GDF-5; LAP4; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; OS5; radotermin; SYM1B; SYNS2 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
8200 | |
Human GDF-5 | |
-20°C | |
<1 EU/μg | |
GDF5 | |
Unconjugated | |
APLATRQGKR PSKNLKARCS RKALHVNFKD MGWDDWIIAP LEYEAFHCEG LCEFPLRSHL EPTNHAVIQT LMNSMDPEST PPTCCVPTRL SPISILFIDS ANNVVYKQYE DMVVESCGCR | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |