missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Heregulin beta-1 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £1200.00
Specifications
Accession Number | Q02297 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3084 |
Molecular Weight (g/mol) | 7.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17830953
|
Gibco™
100-03-100UG |
100 μg |
£240.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870983
|
Gibco™
100-03-10UG |
10 μg |
£64.75
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17851273
|
Gibco™
100-03-1MG |
1 mg |
£1200.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17881353
|
Gibco™
100-03-250UG |
250 μg |
£421.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17821183
|
Gibco™
100-03-500UG |
500 μg |
£720.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17810953
|
Gibco™
100-03-50UG |
50 μg |
£157.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Direct ligand for ERBB3 and ERBB4 tyrosine kinase receptors. Concomitantly recruits ERBB1 and ERBB2 coreceptors, resulting in ligand-stimulated tyrosine phosphorylation and activation of the ERBB receptors. The multiple isoforms perform diverse functions such as inducing growth and differentiation of epithelial, glial, neuronal, and skeletal muscle cells; inducing expression of acetylcholine receptor in synaptic vesicles during the formation of the neuromuscular junction; stimulating lobuloalveolar budding and milk production in the mammary gland and inducing differentiation of mammary tumor cells; stimulating Schwann cell proliferation; implication in the development of the myocardium such as trabeculation of the developing heart. Isoform 10 may play a role in motor and sensory neuron development.Specifications
Q02297 | |
protein with no preservative | |
7.5 | |
E. coli | |
RUO | |
6030402G23Rik; acetylcholine receptor-inducing activity; ARIA; Breast cancer cell differentiation factor p45; D230005F13Rik; GGF; GGF2; GGFII; Glial growth factor; glial growth factor 2; heregulin; heregulin, alpha (45 kD, ERBB2 p185-activator); Hgl; HRG; HRG1; HRGA; HRGalpha; MST131; MSTP131; NDF; Neu differentiation factor; neuregulin 1; neuregulin 1 type III beta 3; Neuregulin-1; Nrg1; NRG1-IT2; Pro-neuregulin-1, membrane-bound isoform; pro-NRG1; sensory and motor neuron derived factor; Sensory and motor neuron-derived factor; SMDF; type I neuregulin 1; type III neuregulin 1 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
3084 | |
Human Heregulin beta-1 | |
-20°C | |
<1 EU/μg | |
Nrg1 | |
Unconjugated | |
SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |