missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human HSP27 Platinum ELISA Kit
For the quantitative detection of human HSP27.
£451.00 - £3070.00
Specifications
Assay Sensitivity | 10.9pg/mL |
---|---|
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
Target Species | Human |
For Use With (Application) | ELISA |
Synonym | Heat shock protein 27, heat shock protein beta-1, germ cell-dependent phosphoprotein-1 |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP27
Biotin-Conjugate anti-human HSP27 monoclonal antibody
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15541607
|
Invitrogen™
BMS2086 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
15571607
|
Invitrogen™
BMS2086TWO |
2 x 96 Tests |
£618.00
Pack of 2 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
15551607
|
Invitrogen™
BMS2086FIVE |
5 x 96 Tests |
£1548.00
Pack of 5 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
15561607
|
Invitrogen™
BMS2086TEN |
10 x 96 Tests |
£3070.00
Pack of 10 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
10.9pg/mL | |
Human | |
Heat shock protein 27, heat shock protein beta-1, germ cell-dependent phosphoprotein-1 |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
ELISA | |
hsp27 |