missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human HSP27 Platinum ELISA Kit

For the quantitative detection of human HSP27.

Brand:  Invitrogen™ BMS2086FIVE

1032.05 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 15551607

  • £1548.00 / Pack of 5
Estimated Shipment: 07-10-2024
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price


Includes:
  • Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP27
  • Biotin-Conjugate anti-human HSP27 monoclonal antibody
This item is not returnable. View return policy

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10.9pg/mL
Human
5 x 96 Tests
hsp27
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
ELISA
Heat shock protein 27, heat shock protein beta-1, germ cell-dependent phosphoprotein-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers