Learn More
Invitrogen™ IgG (Total) Human ELISA Kit
For the quantitative detection of human IgG.
£451.00 - £3070.00
Specifications
Assay Range | 1.6-100 ng/mL |
---|---|
Assay Sensitivity | 0.24 ng/mL |
Conjugate | HRP |
Product Type | ELISA Kit |
Sample Type | Plasma, Serum |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human total IgG
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15581617
|
Invitrogen™
BMS2091 |
96 Tests |
£451.00
96 tests |
In Stock
Log in to see stock. |
|||||
15591617
|
Invitrogen™
BMS2091TEN |
10 x 96 Tests |
£3070.00
Pack of 10 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
Description
The Human Total Immunoglobulin G (Hu IgG) ELISA quantitates Hu IgG in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu IgG. Principle of the method The Human IgG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
1.6-100 ng/mL | |
HRP | |
Plasma, Serum | |
ELISA | |
2.6% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
Human | |
1 hr. 30 min. |
0.24 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
5.3% | |
HRP | |
Plasma, 10 μL; Serum, 10 μL | |
Immunoglobulin G | |
1 hr. 20 min. |
For Research Use Only