missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-17E (IL-25) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 200-24-100UG
This item is not returnable.
View return policy
Description
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.Specifications
Q9H293 | |
protein with no preservative | |
33.8 | |
100 μg | |
-20°C | |
<1 EU/μg | |
IL25 | |
Unconjugated | |
MYSHWPSCCP SKGQDTSEEL LRWSTVPVPP LEPARPNRHP ESCRASEDGP LNSRAISPWR YELDRDLNRL PQDLYHARCL CPHCVSLQTG SHMDPRGNSE LLYHNQTVFY RRPCHGEKGT HKGYCLERRL YRVSLACVCV RPRVMG | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
64806 | |
Human IL-17 E (IL-25) | |
E. coli | |
RUO | |
D17Wsu104e; il 17 e; Il17e; IL-17 E; Il25; IL-25; ILN; Interleukin; interleukin 17 E; interleukin 25; interleukin-17 E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog | |
Human | |
Recombinant | |
Lyophilized |