missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-17E (IL-25) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £2400.00
Specifications
Accession Number | Q9H293 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 64806 |
Molecular Weight (g/mol) | 33.8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17831623
|
Gibco™
200-24-100UG |
100 μg |
£480.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17851613
|
Gibco™
|
2 x 500 μg | N/A | ||||||
17801323
|
Gibco™
200-24-250UG |
250 μg |
£838.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17810443
|
Gibco™
200-24-25UG |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17800243
|
Gibco™
200-24-500UG |
500 μg |
£1436.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17801423
|
Gibco™
200-24-5UG |
5 μg |
£64.75
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.Specifications
Q9H293 | |
protein with no preservative | |
33.8 | |
E. coli | |
RUO | |
D17Wsu104e; il 17 e; Il17e; IL-17 E; Il25; IL-25; ILN; Interleukin; interleukin 17 E; interleukin 25; interleukin-17 E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
64806 | |
Human IL-17 E (IL-25) | |
-20°C | |
<1 EU/μg | |
IL25 | |
Unconjugated | |
MYSHWPSCCP SKGQDTSEEL LRWSTVPVPP LEPARPNRHP ESCRASEDGP LNSRAISPWR YELDRDLNRL PQDLYHARCL CPHCVSLQTG SHMDPRGNSE LLYHNQTVFY RRPCHGEKGT HKGYCLERRL YRVSLACVCV RPRVMG | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |