missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-3 DuoSet ELISA
contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Human IL-3
Brand: R&D Systems™ DY203
Additional Details : Weight : 0.09700kg
Description
The Human IL-3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-3.Specifications
ELISA Development Kit | |
Human | |
15.60 to 1000 pg/mL | |
ELISA | |
Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
3562 | |
15 Plates |