missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-3 DuoSet ELISA
contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Human IL-3
£232.18 - £437.35
Specifications
Product Type | ELISA Development Kit |
---|---|
Quantity | 1 Kit |
Species | Human |
Conjugate | Biotin |
Assay Range | 15.60 to 1000 pg/mL |
Product Code | Brand | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Quantity | Price | Quantity & Availability | |||||
30021740
|
R&D Systems™
DY203 |
15 Plates |
£437.35
1 set |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30021560
|
R&D Systems™
DY203-05 |
5 Plates |
£232.18
1 set |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Human IL-3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-3.Specifications
ELISA Development Kit | |
Human | |
15.60 to 1000 pg/mL | |
ELISA | |
Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
3562 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |