missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-34 Recombinant Protein, PeproTech®
HEK293 cells Recombinant Protein
Brand: Gibco™ 200-34-01M
This item is not returnable.
View return policy
Description
IL-34 is secreted as a dimeric protein and currently belongs to no known cytokine family. It signals via the M-CSF Receptor, also known as CSF-1R and CD115, which is shared with M-CSF. IL-34 has no amino acid sequence homology to M-CSF, but they exhibit similarities in secondary structure and share overlapping functions. IL-34 has been shown to promote myeloid survival, proliferation, and differentiation, and to play a role in osteoclast formation. IL-34 expression has been detected in various tissues and is most abundant in the spleen.Specifications
Q6ZMJ4 | |
Protein with no preservative | |
52.5 | |
2 x 500 μg | |
-20°C | |
<1 EU/μg | |
Il34 | |
Unconjugated | |
NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP KVESVLSLLN APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHHHH | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
146433 | |
Human IL-34 | |
HEK293 cells | |
RUO | |
2010004A03Rik; AI593503; C16orf77; IL34; IL-34; ILN; Interleukin; interleukin 34; Interleukin34; interleukin-34 | |
Human | |
Recombinant | |
Lyophilized |