missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-34 Recombinant Protein, PeproTech®
HEK293 cells Recombinant Protein
£64.75 - £4150.00
Specifications
Accession Number | Q6ZMJ4 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 146433 |
Molecular Weight (g/mol) | 52.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17821883
|
Gibco™
200-34-100UG |
100 μg |
£832.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17850433
|
Gibco™
200-34-10UG |
10 μg |
£157.00
10µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17810063
|
Gibco™
200-34-1MG |
2 x 500 μg |
£4150.00
1mg |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17870433
|
Gibco™
200-34-250UG |
250 μg |
£1456.00
250µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17821813
|
Gibco™
200-34-2UG |
2 μg |
£64.75
2µg |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17850443
|
Gibco™
200-34-500UG |
500 μg |
£2495.00
500µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17871873
|
Gibco™
200-34-50UG |
50 μg |
£502.00
50µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
IL-34 is secreted as a dimeric protein and currently belongs to no known cytokine family. It signals via the M-CSF Receptor, also known as CSF-1R and CD115, which is shared with M-CSF. IL-34 has no amino acid sequence homology to M-CSF, but they exhibit similarities in secondary structure and share overlapping functions. IL-34 has been shown to promote myeloid survival, proliferation, and differentiation, and to play a role in osteoclast formation. IL-34 expression has been detected in various tissues and is most abundant in the spleen.Specifications
Q6ZMJ4 | |
protein with no preservative | |
52.5 | |
HEK293 cells | |
RUO | |
2010004A03Rik; AI593503; C16orf77; IL34; IL-34; ILN; Interleukin; interleukin 34; Interleukin34; interleukin-34 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
146433 | |
Human IL-34 | |
-20°C | |
<1 EU/μg | |
Il34 | |
Unconjugated | |
NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP KVESVLSLLN APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHHHH | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |