missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-7 Recombinant Protein
Recombinant Human Interleukin 7 (IL7) is a bioactive protein intended for use in cell culture applications
£108.00 - £6210.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 17.5 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | IL-7 |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10571085
|
Gibco™
PHC0075 |
5μg |
£108.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10068782
|
Gibco™
PHC0076 |
25μg |
£906.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10416673
|
Gibco™
PHC0071 |
100μg |
£2050.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10140813
|
Gibco™
PHC0073 |
1mg |
£6210.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human IL-7 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL7 stimulates the proliferation of pre-B and pro-B cells. IL7 also supports the maturation of megakaryocytes.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human PBMCs
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Specifications
Cell Culture | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Signal Transduction, Stem Cell Research, Drug Development, Oncology | |
95 % |
17.5 kDa | |
IL-7 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.