missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human iRHOM2/RHBDF2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB10048-100
This item is not returnable.
View return policy
Description
RHBDF2 Monoclonal specifically detects RHBDF2 in Human samples. It is validated for Western Blot.Specifications
RHBDF2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FLJ22341, iRHOM2, RHBDL5, RHBDL5rhomboid family member 2, RHBDL6, Rhomboid 5 homolog 2, rhomboid 5 homolog 2 (Drosophila), rhomboid veinlet-like 5, Rhomboid veinlet-like protein 5, Rhomboid veinlet-like protein 6, rhomboid, veinlet-like 6, rhomboid, veinlet-like 6 (Drosophila), rhomboid-like protein 6, TOCG | |
Synthetic peptide containing human iRHOM2/RHBDF2, Accession No. Q6PJF2 | |
100 μg | |
79651.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
996308 | |
Western Blot 2 μg/mL | |
Q6PJF2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects huma iRHOM2/RHBDF2 in direct ELISAs and Western blots. | |
Human | |
Supernatant |