missing translation for 'onlineSavingsMsg'
Learn More

Human iRHOM2/RHBDF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody

£152.00 - £394.00

Specifications

Antigen RHBDF2
Clone 996308
Dilution Western Blot 2 μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18714637
View Documents
R&D Systems
MAB10048-SP
25 μg
£152.00
25µg
Please to purchase this item. Need a web account? Register with us today!
18732377
View Documents
R&D Systems
MAB10048-100
100 μg
£394.00
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RHBDF2 Monoclonal specifically detects RHBDF2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RHBDF2
Western Blot 2 μg/mL
Monoclonal
Supernatant
Human
Q6PJF2
79651.0
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
996308
Western Blot
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
FLJ22341, iRHOM2, RHBDL5, RHBDL5rhomboid family member 2, RHBDL6, Rhomboid 5 homolog 2, rhomboid 5 homolog 2 (Drosophila), rhomboid veinlet-like 5, Rhomboid veinlet-like protein 5, Rhomboid veinlet-like protein 6, rhomboid, veinlet-like 6, rhomboid, veinlet-like 6 (Drosophila), rhomboid-like protein 6, TOCG
Synthetic peptide containing human iRHOM2/RHBDF2, Accession No. Q6PJF2
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects huma iRHOM2/RHBDF2 in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications