missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LAIR1 ELISA Kit
Human LAIR1 quantitates human LAIR1 in serum, plasma, supernatant.
Brand: Invitrogen™ EH291RB
This item is not returnable.
View return policy
Description
Human LAIR1 quantitates human LAIR1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LAIR1.
LAIR1 (CD305) functions as an inhibitory receptor that plays a constitutive negative regulatory role on cytolytic function of natural killer (NK) cells, B-cells and T-cells. Activation by Tyr phosphorylation results in recruitment and activation of the phosphatases PTPN6 and PTPN11. It also reduces the increase of intracellular calcium evoked by B-cell receptor ligation. May also play its inhibitory role independently of SH2-containing phosphatases. Modulates cytokine production in CD4+ T-cells, down-regulating IL2 and IFNG production while inducing secretion of transforming growth factor beta. Down-regulates also IgG and IgE production in B-cells as well as IL8, IL10 and TNF secretion. Inhibits proliferation and induces apoptosis in myeloid leukemia cell lines as well as prevents nuclear translocation of NF-kappa-B p65 subunit/RELA and phosphorylation of I-kappa-B alpha/CHUK in these cells. Inhibits the differentiation of peripheral blood precursors towards dendritic cells.Specifications
5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; LAIR-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1 | |
0.6 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
LAIR1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.6 to 130 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q6GTX8 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
LAIR1 (CD305) | |
4 hr 45 min |