missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LAIR1 ELISA Kit

Human LAIR1 quantitates human LAIR1 in serum, plasma, supernatant.

£540.00 - £2320.00

Specifications

Accession Number 5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; LAIR-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1
Assay Range 0.6 to 130 ng/mL
Assay Sensitivity 0.6 ng/mL
Conjugate Biotin
Product Type ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17155059
View Documents Promotion Details
Invitrogen™
EH291RBX5
5 x 96 Tests
£2320.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-11-2024
to see stock.
Add to basket
 
17174583
View Documents Promotion Details
Invitrogen™
EH291RB
96 Tests
£540.00
Pack of 96
View alternative products
Save Up to 
»
Estimated Shipment: 05-11-2024
to see stock.
Add to basket
 
Description

Description

Human LAIR1 quantitates human LAIR1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LAIR1.

LAIR1 (CD305) functions as an inhibitory receptor that plays a constitutive negative regulatory role on cytolytic function of natural killer (NK) cells, B-cells and T-cells. Activation by Tyr phosphorylation results in recruitment and activation of the phosphatases PTPN6 and PTPN11. It also reduces the increase of intracellular calcium evoked by B-cell receptor ligation. May also play its inhibitory role independently of SH2-containing phosphatases. Modulates cytokine production in CD4+ T-cells, down-regulating IL2 and IFNG production while inducing secretion of transforming growth factor beta. Down-regulates also IgG and IgE production in B-cells as well as IL8, IL10 and TNF secretion. Inhibits proliferation and induces apoptosis in myeloid leukemia cell lines as well as prevents nuclear translocation of NF-kappa-B p65 subunit/RELA and phosphorylation of I-kappa-B alpha/CHUK in these cells. Inhibits the differentiation of peripheral blood precursors towards dendritic cells.
Specifications

Specifications

5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; LAIR-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1
0.6 ng/mL
ELISA
Colorimetric Microplate Reader
LAIR1
<10%
HRP
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL
LAIR1 (CD305)
4 hr 45 min
0.6 to 130 ng/mL
Biotin
Plasma, Serum, Supernatant
Q6GTX8
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Research Use Only
2°C to 8°C
1 hr 20 min
Videos
SDS
Documents

Documents

Product Certifications