missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human MD-2/LY96 Recombinant Protein, PeproTech®
HEK293 cells Recombinant Protein
Brand: Gibco™ 160-07-10UG
58.50 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Toll-like receptors (TLRs) are evolutionarily conserved patternrecognition molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. The signaling of these TLRs is kept under tight control by the expression of endogenous inhibiting proteins such as RP105, a recently identified homolog of TLR4. This protein, in association with MD-1, interacts with and inhibits the TLR4/MD-2 signaling pathway. While MD-2 can directly bind to LPS, one of the activating molecules of TLR4, the function of MD -1 is less well-known. It has been suggested however, that the RP105/MD-1 complex influences antibody production mediated by both TLR4/MD-2 and TLR2 receptor complexes.Specifications
Q9Y6Y9 | |
protein with no preservative | |
17.2 | |
10 μg | |
-20°C | |
<1 EU/μg | |
LY96 | |
Unconjugated | |
QKQYWVCNSS DASISYTYCD KMQYPISINV NPCIELKGSK GLLHIFYIPR RDLKQLYFNL YITVNTMNLP KRKEVICRGS DDDYSFCRAL KGETVNTTIS FSFKGIKFSK GKYKCVVEAI SGSPEEMLFC LEFVILHQPN SNHHHHHH | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay | |
23643 | |
Human MD-2/LY96 | |
HEK293 cells | |
RUO | |
Esop1; ESOP-1; LPS co-receptor MD-2; LY96; ly-96; Lymphocyte antigen 96; MD2; MD-2; myeloid differentiation factor-2; myeloid differentiation protein-2; protein MD-2 | |
Human | |
Recombinant | |
Lyophilized |