Learn More
Invitrogen™ Human MRP1 Vesicles
Human MRP1 Vesicles are insect derived purified plasma membranes with inserted MRP1 transport protein (multidrug resistance-associated protein 1, ABCC1). Our Human MRP1 Vesicles can be used to evaluate test compounds and drug interactions with MRP1 in in vitro assays.
Brand:  Invitrogen™ GM0010
Product Code. 10212420
  • £680.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Specifications
Frozen
Research Grade
Human Vesicle
Vial
0.5 mL
Human
Store at -80°C.
Dry Ice
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberXB6YD03 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberGM0010
Lot NumberBUDCG03 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberGM0010
Lot NumberIFURG30 Certificate TypeCertificate of Analysis Date31/01/2020 Catalog NumberGM0010
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Human MRP1 Vesicles > MRP1 vesicles