missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human OTOR Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 130-03-250UG
This item is not returnable.
View return policy
Description
OTOR, also called Otoraplin and MIAL, is a secreted cytokine of the MIA/OTOR family. Members of this family, which include MIA, MIA2, and TANGO, share a Src homology-3 (SH3)-like domain. OTOR is predominantly expressed in the cochlea of the inner ear, and, to a lesser extent, in fetal brain tissue and some cartilage tissues. OTOR appears to be involved in early chondrogenesis of the otic capsule, which is required for normal inner ear development and auditory function.Specifications
Q9NRC9 | |
protein with no preservative | |
12.7 | |
250 μg | |
-20°C | |
<1 EU/μg | |
OTOR | |
Unconjugated | |
MVHGIFMDRL ASKKLCADDE CVYTISLASA QEDYNAPDCR FINVKKGQQI YVYSKLVKENGAGEFWAGSV YGDGQDEMGV VGYFPRNLVK EQRVYQEATK EVPTTDIDFF CE | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
56914 | |
Human OTOR | |
E. coli | |
RUO | |
CDRAP; FDP; Fibrocyte-derived protein; melanoma inhibitory activity-like protein; MIA; MIAL; MIAL1; Otor; Otoraplin; UNQ3054/PRO9873 | |
Human | |
Recombinant | |
Lyophilized |