missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human OTOR Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £3125.00
Specifications
Accession Number | Q9NRC9 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 56914 |
Molecular Weight (g/mol) | 12.7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17880753
|
Gibco™
130-03-100UG |
100 μg |
£624.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830663
|
Gibco™
130-03-1MG |
1 mg |
£3125.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861733
|
Gibco™
130-03-20UG |
20 μg |
£158.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17831143
|
Gibco™
130-03-250UG |
250 μg |
£1094.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830553
|
Gibco™
130-03-500UG |
500 μg |
£1876.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17851343
|
Gibco™
130-03-5UG |
5 μg |
£65.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OTOR, also called Otoraplin and MIAL, is a secreted cytokine of the MIA/OTOR family. Members of this family, which include MIA, MIA2, and TANGO, share a Src homology-3 (SH3)-like domain. OTOR is predominantly expressed in the cochlea of the inner ear, and, to a lesser extent, in fetal brain tissue and some cartilage tissues. OTOR appears to be involved in early chondrogenesis of the otic capsule, which is required for normal inner ear development and auditory function.Specifications
Q9NRC9 | |
protein with no preservative | |
12.7 | |
E. coli | |
RUO | |
CDRAP; FDP; Fibrocyte-derived protein; melanoma inhibitory activity-like protein; MIA; MIAL; MIAL1; Otor; Otoraplin; UNQ3054/PRO9873 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
56914 | |
Human OTOR | |
-20°C | |
<1 EU/μg | |
OTOR | |
Unconjugated | |
MVHGIFMDRL ASKKLCADDE CVYTISLASA QEDYNAPDCR FINVKKGQQI YVYSKLVKENGAGEFWAGSV YGDGQDEMGV VGYFPRNLVK EQRVYQEATK EVPTTDIDFF CE | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |