missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WDR32 Partial ORF (NP_077321, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079269-Q01.25ug
This item is not returnable.
View return policy
Description
Sequence: MTSLYGSIHPADSVYLSTRTHGAVFNLEYSPDGSVLTVACEQTEVLLFDPISSKHIKTLSEAHEDCVNNIRFLDNRLFATCSDDTTIALWDLRKLNTKVCTLHGHTSWVKSpecifications
NP_077321 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTSLYGSIHPADSVYLSTRTHGAVFNLEYSPDGSVLTVACEQTEVLLFDPISSKHIKTLSEAHEDCVNNIRFLDNRLFATCSDDTTIALWDLRKLNTKVCTLHGHTSWVK | |
RUO | |
WDR32 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
79269 | |
WDR32 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23201/MGC10765/MGC131781 | |
WDR32 | |
Recombinant | |
wheat germ expression system |