missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WDR32 Partial ORF (NP_077321, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
£280.00 - £425.00
Specifications
Accession Number | NP_077321 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79269 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16169986
|
Abnova™
H00079269-Q01.10ug |
10 ug |
£280.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16179986
|
Abnova™
H00079269-Q01.25ug |
25 ug |
£425.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence: MTSLYGSIHPADSVYLSTRTHGAVFNLEYSPDGSVLTVACEQTEVLLFDPISSKHIKTLSEAHEDCVNNIRFLDNRLFATCSDDTTIALWDLRKLNTKVCTLHGHTSWVKSpecifications
NP_077321 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23201/MGC10765/MGC131781 | |
WDR32 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79269 | |
WDR32 (Human) Recombinant Protein (Q01) | |
MTSLYGSIHPADSVYLSTRTHGAVFNLEYSPDGSVLTVACEQTEVLLFDPISSKHIKTLSEAHEDCVNNIRFLDNRLFATCSDDTTIALWDLRKLNTKVCTLHGHTSWVK | |
RUO | |
WDR32 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |