Learn More
UVP Hybridization Oven Bottles and Accessories
Brand:  UVP 76-0089-01
Code : 04
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 12893396
  • £367.00 / Each
Estimated Shipment: 18-09-2024
to see stock.
Add to basket
Specifications
Polypropylene, PTFE
DNA, RNA, or protein identification
UVP hybridization ovens
Acrylic Carousel (for upper chamber)
Borosilicate Glass
150 or 300 mm bottles