Learn More
Cytiva HyClone™ ActiCHO™ SM Powdered Media
Hydrolysate-free and animal-derived component-free (ADCF)
Brand: Cytiva SH31029.01
Additional Details : CAS Number : 56-45-1 Weight : 0.12810kg
Description
ActiCHO SM powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.
- Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides or growth factors in ActiCHO SM medium.
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
No Glutamine, No Insulin |
Powder | |
Poloxamer 188 | |
5 L |