missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ ActiCHO™ SM Powdered Media
Hydrolysate-free and animal-derived component-free (ADCF)
£73.90 - £296.00
Specifications
Content And Storage | 2 to 8°C, away from light and moisture |
---|---|
Form | Powder |
Sterility | Non-sterile |
With Additives | Poloxamer 188 |
Without Additives | No Glutamine, No Insulin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17275896
|
Cytiva
SH31029.06 |
1 L |
£73.90
1 L |
Estimated Shipment: 30-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17265896
|
Cytiva
SH31029.01 |
5 L |
£296.00
5 L |
Estimated Shipment: 30-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ActiCHO SM powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.
- Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides or growth factors in ActiCHO SM medium.
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
No Glutamine, No Insulin |
Powder | |
Poloxamer 188 |