missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ AdvanceSTEM™ Dulbecco's Phosphate Buffered Saline, ES Cell Qualified
Brand: Cytiva SH30850.03
Code : 85
Additional Details : Weight : 1.05000kg
Description
Support growth of embryonic cell culture growth with HyClone™ AdvanceSTEM™ Dulbecco's Phosphate Buffered Saline.
Specifications
cGMP and ISO 91:2 Guidelines | |
AdvanceSTEM ES Qualified DPBS without Calcium or Magnesium, 1000mL | |
0.1 μm Sterile-filtered | |
=1.0EU/mL | |
1000 mL |
15 to 30°C | |
1 X | |
No Calcium, No Magnesium | |
7.0 to 7.2 |