missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ AdvanceSTEM™ Dulbecco's Phosphate Buffered Saline, ES Cell Qualified
£20.85 - £22.05
Specifications
Certifications/Compliance | cGMP and ISO 91:2 Guidelines |
---|---|
Content And Storage | 15 to 30°C |
Concentration | 1 X |
Sterilization Method | 0.1 μm Sterile-filtered |
Without Additives | No Calcium, No Magnesium |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12310103
|
Cytiva
SH30850.02 |
AdvanceSTEM ES Qualified DPBS without Calcium or Magnesium, 500mL |
£20.85
500mL |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11526331
|
Cytiva
SH30850.03 |
AdvanceSTEM ES Qualified DPBS without Calcium or Magnesium, 1000mL |
£22.05
1000mL |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Support growth of embryonic cell culture growth with HyClone™ AdvanceSTEM™ Dulbecco's Phosphate Buffered Saline.
Specifications
cGMP and ISO 91:2 Guidelines | |
1 X | |
No Calcium, No Magnesium | |
7.0 to 7.2 |
15 to 30°C | |
0.1 μm Sterile-filtered | |
=1.0EU/mL |